• USA Home
  • 559458 - 3,4,5-Trifluorobenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
559458 Sigma-Aldrich

3,4,5-Trifluorobenzyl alcohol

95%

Purchase

Properties

Related Categories Alcohols, Building Blocks, C7 to C8, Chemical Synthesis, Organic Building Blocks,
InChI Key   HRSFRSLKOPFWMZ-UHFFFAOYSA-N
assay   95%
refractive index   n20/D 1.4700(lit.)
bp   203-204 °C/760 mmHg(lit.)
density   1.385 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?