• USA Home
  • 559539 - 2-Fluoro-5-methylanisole

EMAIL THIS PAGE TO A FRIEND
559539 Sigma-Aldrich

2-Fluoro-5-methylanisole

97%

  • CAS Number 63762-78-7

  • Linear Formula FC6H3(CH3)OCH3

  • Molecular Weight 140.15

  •  MDL number MFCD00665791

  •  PubChem Substance ID 24879620

  •  NACRES NA.22

Purchase

Properties

Related Categories Aryl Fluorinated Building Blocks, Building Blocks, C8, C8-C12, Chemical Synthesis,
InChI Key   MOFZLPPSEOEXNJ-UHFFFAOYSA-N
Quality Level   100
assay   97%
refractive index   n20/D 1.4950 (lit.)
bp   176-177 °C (lit.)
density   1.070 g/mL at 25 °C (lit.)

Description

General description

2-Fluoro-5-methylanisole (FMA) undergoes" copolymerization" with other monomers and contains the nanoparticles, to form hydrogels useful for ophthalmic lenses.

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
172.4 °F
Flash Point(C) 
78 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?