• USA Home
  • 559776 - 3-(Trifluoromethyl)-4-methylphenyl isocyanate

EMAIL THIS PAGE TO A FRIEND
559776 Sigma-Aldrich

3-(Trifluoromethyl)-4-methylphenyl isocyanate

97%

Purchase

Properties

assay   97%
refractive index   n20/D 1.4800 (lit.)
bp   190-191 °C (lit.)
density   1.288 g/mL at 25 °C (lit.)
SMILES string   Cc1ccc(cc1C(F)(F)F)N=C=O
InChI   1S/C9H6F3NO/c1-6-2-3-7(13-5-14)4-8(6)9(10,11)12/h2-4H,1H3
InChI key   XWSZWQGFJRBXMO-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 2206PSN1B 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
174.2 °F
Flash Point(C) 
79 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?