• USA Home
  • 560634 - Bis(cyclopentadienyl)hafnium(IV) dihydride

EMAIL THIS PAGE TO A FRIEND
560634 Sigma-Aldrich

Bis(cyclopentadienyl)hafnium(IV) dihydride

Synonym: Bis(cyclopentadienyl)dihydridohafnium(IV), Hafnocene dihydride

Purchase

Properties

mp   226 °C (dec.) (lit.)
SMILES string   [HfH2].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3131 8(4.3) / PGII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?