• USA Home
  • 560782 - 2-Methoxy-5-(3′,7′-dimethyloctyloxy)terephthalaldehyde

EMAIL THIS PAGE TO A FRIEND
560782 Sigma-Aldrich

2-Methoxy-5-(3′,7′-dimethyloctyloxy)terephthalaldehyde

98%

  • CAS Number 207394-56-7

  • Linear Formula (CH3)2CH(CH2)3CH(CH3)CH2CH2OC6H2(CH=O)2OCH3

  • Molecular Weight 320.42

  •  MDL number MFCD03427238

  •  PubChem Substance ID 24879893

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C13-C60, Carbonyl Compounds, Chemical Synthesis,
assay   98%
mp   81.0-86.8 °C (lit.)
SMILES string   [H]C(=O)c1cc(OCCC(C)CCCC(C)C)c(cc1OC)C([H])=O
InChI   1S/C19H28O4/c1-14(2)6-5-7-15(3)8-9-23-19-11-16(12-20)18(22-4)10-17(19)13-21/h10-15H,5-9H2,1-4H3
InChI key   IPIUNLMDSXJBFQ-UHFFFAOYSA-N

Description

General description

Monomeric precursor for the light emitting polymer OC1C10-CN-PPV

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?