• USA Home
  • 562394 - Triethylphosphine oxide

EMAIL THIS PAGE TO A FRIEND
562394 Sigma-Aldrich

Triethylphosphine oxide

97%

  • CAS Number 597-50-2

  • Linear Formula (C2H5)3PO

  • Molecular Weight 134.16

  •  MDL number MFCD00013914

  •  PubChem Substance ID 24880030

  •  NACRES NA.22

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Phosphine Compounds, Phosphine Ligands
assay   97%
mp   48-52 °C (lit.)
functional group   phosphine
reaction suitability   reaction type: Buchwald-Hartwig Cross Coupling Reaction
  reaction type: Heck Reaction
  reaction type: Hiyama Coupling
  reaction type: Negishi Coupling
  reaction type: Sonogashira Coupling
  reaction type: Stille Coupling
  reaction type: Suzuki-Miyaura Coupling
  reagent type: ligand
SMILES string   CCP(=O)(CC)CC
InChI   1S/C6H15OP/c1-4-8(7,5-2)6-3/h4-6H2,1-3H3
InChI key   ZSSWXNPRLJLCDU-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
SZ1640000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?