• USA Home
  • 56244 - trans-3-Hydroxy-L-proline

EMAIL THIS PAGE TO A FRIEND
56244 Aldrich

trans-3-Hydroxy-L-proline

≥98.0% (NT)

Synonym: (2S,3S)-3-Hydroxypyrrolidine-2-carboxylic acid

  • CAS Number 4298-08-2

  • Empirical Formula (Hill Notation) C5H9NO3

  • Molecular Weight 131.13

  •  Beilstein Registry Number 471959

  •  MDL number MFCD00216471

  •  PubChem Substance ID 57651573

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry | Peptide Synthesis, Proline Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   BJBUEDPLEOHJGE-IMJSIDKUSA-N
assay   ≥98.0% (NT)
optical activity   [α]20/D −17.5±2°, c = 4% in H2O

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Articles

Proline Derivatives and Analogs

Proline is a non-polar proteinogenic amino acid that forms a tertiary amide when incorporated into peptides. It does not have a hydrogen on the amide group and therefore cannot act as a hydrogen bond...
Chemfiles Volume 5 Article 12
Keywords: Adhesion, Biochemistry, Catalysis, Deposition, Hydroxylations, Isomerizations, Nucleic acid hybridization, Pharmaceutical, Reductions, Substitutions, Type

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?