• USA Home
  • 563293 - 4-Isopropylphenylacetonitrile

EMAIL THIS PAGE TO A FRIEND
563293 Sigma-Aldrich

4-Isopropylphenylacetonitrile

99%

Purchase

Properties

assay   99%
refractive index   n20/D 1.5125 (lit.)
bp   85-90 °C/0.2 atm (lit.)
density   0.960 g/mL at 25 °C (lit.)
SMILES string   CC(C)c1ccc(CC#N)cc1
InChI   1S/C11H13N/c1-9(2)11-5-3-10(4-6-11)7-8-12/h3-6,9H,7H2,1-2H3
InChI key   RIPHZOPMCRSGSI-UHFFFAOYSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?