• USA Home
  • 563323 - Cyclohexane-1,4-dicarbonitrile

EMAIL THIS PAGE TO A FRIEND
563323 Sigma-Aldrich

Cyclohexane-1,4-dicarbonitrile

98% Mixture of cis and trans

Purchase

Properties

Related Categories Building Blocks, C8 to C9, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds - Organic Building Blocks,
Quality Level   100
mp   57-61 °C (lit.)
SMILES string   N#CC1CCC(CC1)C#N
InChI   1S/C8H10N2/c9-5-7-1-2-8(6-10)4-3-7/h7-8H,1-4H2
InChI key   MGWYSXZGBRHJNE-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?