• USA Home
  • 563668 - 2-(4-Pentynyloxy)tetrahydro-2H-pyran

EMAIL THIS PAGE TO A FRIEND
563668 Sigma-Aldrich

2-(4-Pentynyloxy)tetrahydro-2H-pyran

97%

  • CAS Number 62992-46-5

  • Linear Formula C5H9OOCH2CH2CH2C≡CH

  • Molecular Weight 168.23

  •  MDL number MFCD00792655

  •  PubChem Substance ID 24880119

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Pyrans
InChI Key   TZRUKFFDSQQSCK-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.4570 (lit.)
bp   40-45 °C/0.03 mmHg (lit.)
mp   84-88 °C (lit.)
density   0.968 g/mL at 25 °C (lit.)
storage temp.   2-8°C

Description

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
176 °F
Flash Point(C) 
80 °C
Protocols & Articles

Related Content

Alkynes and Acetylenic Building Blocks

We list over 600 alkyne building blocks, including a wide array of propargyl alcohols, acetylenic boron reagents, and halogenated substrates for cross-coupling. Additionally, many of these are termin...
Keywords: Building blocks, Coupling reactions, Cross couplings

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?