• USA Home
  • 566128 - Indenylhafnium(IV) Trichloride

EMAIL THIS PAGE TO A FRIEND
566128 Sigma-Aldrich

Indenylhafnium(IV) Trichloride

98%

  • CAS Number 336102-54-6

  • Empirical Formula (Hill Notation) C9H7Cl3Hf

  • Molecular Weight 400.00

  •  MDL number MFCD03427277

  •  PubChem Substance ID 24880279

  •  NACRES NA.22

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Hafnium Catalysts More...
assay   98%
mp   164-166 °C (lit.)
reaction suitability   reagent type: catalyst
SMILES string   Cl[Hf](Cl)Cl.[CH]1[CH][C]2C=CC=C[C]2[CH]1
InChI   1S/C9H7.3ClH.Hf/c1-2-5-9-7-3-6-8(9)4-1;;;;/h1-7H;3*1H;/q;;;;+3/p-3
InChI key   TXMSRHVGGMAXBL-UHFFFAOYSA-K
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?