• USA Home
  • 566314 - 1-(2-Fluorophenyl)biguanide hydrochloride

EMAIL THIS PAGE TO A FRIEND
566314 Sigma-Aldrich

1-(2-Fluorophenyl)biguanide hydrochloride

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Guanidines, Nitrogen Compounds - Organic Building Blocks, Organic Building Blocks More...
assay   97%
mp   237-241 °C (lit.)
SMILES string   Cl[H].NC(=N)NC(=N)Nc1ccccc1F
InChI   1S/C8H10FN5.ClH/c9-5-3-1-2-4-6(5)13-8(12)14-7(10)11;/h1-4H,(H6,10,11,12,13,14);1H
InChI key   YUQPLHJDRXWUON-UHFFFAOYSA-N

Description

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?