• USA Home
  • 566489 - 1-(2,5-Difluorophenyl)biguanide hydrochloride

EMAIL THIS PAGE TO A FRIEND
566489 Sigma-Aldrich

1-(2,5-Difluorophenyl)biguanide hydrochloride

  • CAS Number 66088-53-7

  • Linear Formula C6H3F2NHC(=NH)NHC(=NH)NH2 · HCl

  • Molecular Weight 249.65

  •  MDL number MFCD03094448

  •  PubChem Substance ID 24880300

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Guanidines, Nitrogen Compounds - Organic Building Blocks, Organic Building Blocks More...
mp   225-230 °C (lit.)
SMILES string   Cl[H].NC(=N)NC(=N)Nc1cc(F)ccc1F
InChI   1S/C8H9F2N5.ClH/c9-4-1-2-5(10)6(3-4)14-8(13)15-7(11)12;/h1-3H,(H6,11,12,13,14,15);1H
InChI key   VVAARBLQYHGNAY-UHFFFAOYSA-N

Description

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?