• USA Home
  • 566772 - Bis(cyclopentadienyl)vanadium(III) chloride

EMAIL THIS PAGE TO A FRIEND
566772 Sigma-Aldrich

Bis(cyclopentadienyl)vanadium(III) chloride

97%

Purchase

Properties

assay   97%
mp   182-194 °C (dec.) (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN3285 - class 6.1 - PG 3 - MP - Vanadium compound, n.o.s., HI: all (not BR)
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?