• USA Home
  • 567698 - 3-Chloro-4-fluorophenylmagnesium bromide solution

EMAIL THIS PAGE TO A FRIEND
567698 Sigma-Aldrich

3-Chloro-4-fluorophenylmagnesium bromide solution

0.5 M in THF

  • CAS Number 413589-34-1

  • Empirical Formula (Hill Notation) C6H3BrClFMg

  • Molecular Weight 233.75

  •  MDL number MFCD02260212

Purchase

Properties

concentration   0.5 M in THF
SMILES string   Fc1ccc([Mg]Br)cc1Cl
InChI   1S/C6H3ClF.BrH.Mg/c7-5-3-1-2-4-6(5)8;;/h2-4H;1H;/q;;+1/p-1
InChI key   XRNCDMQVKMYTAM-UHFFFAOYSA-M
Safety & Documentation

Safety Information

RIDADR 
UN 2924 3/PG 2
WGK Germany 
1
Flash Point(F) 
1 °F
Flash Point(C) 
-17 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

563676 3-Chloro-4-fluorophenylmagnesium bromide solution, 0.5 M in THF

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?