• USA Home
  • 568090 - (1R,2S)-(−)-2-Benzamidocyclohexanecarboxylic acid

EMAIL THIS PAGE TO A FRIEND
568090 Sigma-Aldrich

(1R,2S)-(−)-2-Benzamidocyclohexanecarboxylic acid

97%

Synonym: (1R,2S)-2-(Benzoylamino)cyclohexanecarboxylic acid

  • CAS Number 26693-55-0

  • Empirical Formula (Hill Notation) C14H17 NO3

  • Molecular Weight 247.29

  •  MDL number MFCD00078309

  •  PubChem Substance ID 24880418

Purchase

Properties

assay   97%
optical activity   [α]20/D -34°, c = 1% in chloroform
mp   205-209 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?