• USA Home
  • 56918 - Boc-3-(2-quinolyl)-DL-Ala-OH

EMAIL THIS PAGE TO A FRIEND
56918 Sigma-Aldrich

Boc-3-(2-quinolyl)-DL-Ala-OH

≥95.0%

Synonym: Boc-3-(2-quinolyl)-DL-alanine

Purchase

Properties

Related Categories Alanine Derivatives, Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
assay   ≥95.0%
application(s)   peptide synthesis: suitable
SMILES string   CC(C)(C)OC(=O)NC(Cc1ccc2ccccc2n1)C(O)=O
InChI   1S/C17H20N2O4/c1-17(2,3)23-16(22)19-14(15(20)21)10-12-9-8-11-6-4-5-7-13(11)18-12/h4-9,14H,10H2,1-3H3,(H,19,22)(H,20,21)
InChI key   IKKVPSHCOQHAMU-UHFFFAOYSA-N

Description

Packaging

1 g in poly tube

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?