• USA Home
  • 571547 - Potassium 4-bromophenyltrifluoroborate

EMAIL THIS PAGE TO A FRIEND
571547 Sigma-Aldrich

Potassium 4-bromophenyltrifluoroborate

Purchase

Properties

Related Categories Aryl Trifluoroborate Salts, Boronic Acids and Derivatives, Chemical Synthesis, Organometallic Reagents, Trifluoroborate Salts More...
mp   >300 °C (lit.)
SMILES string   [K+].F[B-](F)(F)c1ccc(Br)cc1
InChI   1S/C6H4BBrF3.K/c8-6-3-1-5(2-4-6)7(9,10)11;/h1-4H;/q-1;+1
InChI key   YTSKPQQWISZEQO-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Chemfiles Volume 4 Article 2 - Potassium Trifluoroborates

Alkyl, aryl, and alkenyl trifluoroborate salts are compelling alternatives to boronic acids in Suzuki–Miyaura coupling and in rhodium catalyzed carbon-carbon bond forming reactions. These salts are a...
Chemfiles Volume 4 Article 2
Keywords: Catalysis, Cross couplings, Ligands

Stabilized 2-Iodoxybenzoic Acid (SIBX)

Since 1994,1 2-iodoxybenzoic acid (IBX) has been well recognized as a very powerful and selective oxidizing agent. Similar to the Dess–Martin periodinane, IBX is an environmentally benign alternative...
ChemFiles 2006, 6.2, 14.
Keywords: Dearomatizations, Oxidations

Related Content

Organotrifluoroborates in Chemical Synthesis

Potent Boronic Acid Surrogates In the vast array of catalytic carbon-carbon bond forming reactions, the Suzuki-Miyaura protocol has proven to be a particularly attractive method.  The organoborane nu...
Keywords: Addition reactions, Catalysis, Coupling reactions, Cross couplings, Halogenations, Suzuki-Miyaura coupling

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?