• USA Home
  • 571679 - 2,5-Dimethoxyphenethyl isocyanate

EMAIL THIS PAGE TO A FRIEND
571679 Sigma-Aldrich

2,5-Dimethoxyphenethyl isocyanate

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Isocyanates, Nitrogen Compounds - Organic Building Blocks, Organic Building Blocks More...
assay   97%
refractive index   n20/D 1.5300 (lit.)
bp   301-302 °C (lit.)
density   1.153 g/mL at 25 °C (lit.)
SMILES string   COc1ccc(OC)c(CCN=C=O)c1
InChI   1S/C11H13NO3/c1-14-10-3-4-11(15-2)9(7-10)5-6-12-8-13/h3-4,7H,5-6H2,1-2H3
InChI key   QUOSISUJAVFTPN-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 2206PSN1B 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?