• USA Home
  • 572225 - 1-(3-Chlorophenyl)biguanide hydrochloride

EMAIL THIS PAGE TO A FRIEND
572225 Sigma-Aldrich

1-(3-Chlorophenyl)biguanide hydrochloride

97%

Synonym: 1-(m-Chlorophenyl)biguanide hydrochloride, N-(3-Chlorophenyl)imidodicarbonimidic diamide hydrochloride, m-CPBG hydrochloride

  • CAS Number 2113-05-5

  • Linear Formula ClC6H4NHC(NH)NHC(NH)NH2 · HCl

  • Molecular Weight 248.11

  •  MDL number MFCD00053019

  •  PubChem Substance ID 24874788

Purchase

Properties

assay   97%
mp   190-194 °C (lit.)
SMILES string   Cl[H].NC(=N)NC(=N)Nc1cccc(Cl)c1
InChI   1S/C8H10ClN5.ClH/c9-5-2-1-3-6(4-5)13-8(12)14-7(10)11;/h1-4H,(H6,10,11,12,13,14);1H
InChI key   FOWAIJYHRWFTHR-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DU1780000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?