• USA Home
  • 572268 - 1-(3,4-Dichlorophenyl)biguanide hydrochloride

EMAIL THIS PAGE TO A FRIEND
572268 Sigma-Aldrich

1-(3,4-Dichlorophenyl)biguanide hydrochloride

97%

  • CAS Number 21703-08-2

  • Linear Formula Cl2C6H3NHC(NH)NHC(NH)NH2 · HCl

  • Molecular Weight 282.56

  •  MDL number MFCD00173791

  •  PubChem Substance ID 24874792

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Guanidines, Nitrogen Compounds - Organic Building Blocks, Organic Building Blocks More...
assay   97%
mp   222-228 °C (lit.)
SMILES string   Cl[H].NC(=N)NC(=N)Nc1ccc(Cl)c(Cl)c1
InChI   1S/C8H9Cl2N5.ClH/c9-5-2-1-4(3-6(5)10)14-8(13)15-7(11)12;/h1-3H,(H6,11,12,13,14,15);1H
InChI key   AMMSXYWPWHREDJ-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?