• USA Home
  • 572454 - Dimethyl 2-fluoroterephthalate

EMAIL THIS PAGE TO A FRIEND
572454 Sigma-Aldrich

Dimethyl 2-fluoroterephthalate

96%

Purchase

Properties

assay   96%
bp   80-90 °C/0.35 mmHg (lit.)
mp   83-85 °C (lit.)
SMILES string   COC(=O)c1ccc(c(F)c1)C(=O)OC
InChI   1S/C10H9FO4/c1-14-9(12)6-3-4-7(8(11)5-6)10(13)15-2/h3-5H,1-2H3
InChI key   NCRFSIQSCLJDIC-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?