• USA Home
  • 573434 - 3-Amino-3-(3-methoxyphenyl)propionic acid

EMAIL THIS PAGE TO A FRIEND
573434 Sigma-Aldrich

3-Amino-3-(3-methoxyphenyl)propionic acid

97%

Purchase

Properties

assay   97%
application(s)   peptide synthesis: suitable
mp   206-210 °C (dec.)
SMILES string   COc1cccc(c1)C(N)CC(O)=O
InChI   1S/C10H13NO3/c1-14-8-4-2-3-7(5-8)9(11)6-10(12)13/h2-5,9H,6,11H2,1H3,(H,12,13)
InChI key   FGMPGCPZEOXEES-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?