• USA Home
  • 573922 - 1,1,1,3,5,7,7,7-Octamethyl-3,5-bis(trimethylsilanyloxy)tetrasiloxane

EMAIL THIS PAGE TO A FRIEND
573922 Sigma-Aldrich

1,1,1,3,5,7,7,7-Octamethyl-3,5-bis(trimethylsilanyloxy)tetrasiloxane

98%

Purchase

Properties

Related Categories Chemical Synthesis, Organometallic Reagents, Organosilicon, Siloxanes
assay   98%
bp   121 °C/10 mmHg (lit.)
density   0.885 g/mL at 25 °C (lit.)
SMILES string   C[Si](C)(C)O[Si](C)(O[Si](C)(C)C)O[Si](C)(O[Si](C)(C)C)O[Si](C)(C)C
InChI   1S/C14H42O5Si6/c1-20(2,3)15-24(13,16-21(4,5)6)19-25(14,17-22(7,8)9)18-23(10,11)12/h1-14H3
InChI key   PMBZCSBOBLHPSR-UHFFFAOYSA-N

Description

Packaging

25 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?