• USA Home
  • 574015 - Cerium(III) oxalate hydrate

EMAIL THIS PAGE TO A FRIEND
574015 Sigma-Aldrich

Cerium(III) oxalate hydrate

99.999% trace metals basis

Synonym: Cerium oxalate nonahydrate

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Ce, Cerium Catalysts, Cerium Salts, Chemical Synthesis,
assay   99.999% trace metals basis
form   powder
SMILES string   O.O=C(O[Ce]1OC(=O)C(=O)O1)C(=O)O[Ce]2OC(=O)C(=O)O2
InChI   1S/3C2H2O4.2Ce.H2O/c3*3-1(4)2(5)6;;;/h3*(H,3,4)(H,5,6);;;1H2/q;;;2*+3;/p-6
InChI key   WSVMKOQJZBJDJB-UHFFFAOYSA-H
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Single- & multi-channel pipettes from BRAND
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?