• USA Home
  • 574104 - Mercury(II) sulfide red

EMAIL THIS PAGE TO A FRIEND
574104 Aldrich

Mercury(II) sulfide red

99.999% trace metals basis

Synonym: Mercuric sulfide red

Purchase

Properties

InChI Key   QXKXDIKCIPXUPL-UHFFFAOYSA-N
assay   99.999% trace metals basis
mp   583.5 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Contact with acids liberates toxic gas.
WGK Germany 
3
RTECS 
OX0720000
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?