• USA Home
  • 574260 - Tetraethylammonium tetrachloromanganate(II)

EMAIL THIS PAGE TO A FRIEND
574260 Sigma-Aldrich

Tetraethylammonium tetrachloromanganate(II)

99.999% trace metals basis

  • CAS Number 6667-73-8

  • Empirical Formula (Hill Notation) C16H40Cl4MnN2

  • Molecular Weight 457.25

  •  MDL number MFCD00075497

  •  PubChem Substance ID 24880721

Purchase

Properties

assay   99.999% trace metals basis
form   powder
mp   >300 °C (lit.)
SMILES string   Cl[Mn--](Cl)(Cl)Cl.CC[N+](CC)(CC)CC.CC[N+](CC)(CC)CC
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?