• USA Home
  • 574457 - 4-(2-Thienyl)-3-buten-2-one

EMAIL THIS PAGE TO A FRIEND
574457 Sigma-Aldrich

4-(2-Thienyl)-3-buten-2-one

97%

Synonym: 1-(2-Thienyl)-1-buten-3-one

Purchase

Properties

Related Categories Building Blocks, C7 to C9, Chemical Synthesis, Heterocyclic Building Blocks, Thiophenes More...
assay   97%
mp   24-29 °C (lit.)
SMILES string   CC(=O)\C=C\c1cccs1
InChI   1S/C8H8OS/c1-7(9)4-5-8-3-2-6-10-8/h2-6H,1H3/b5-4+
InChI key   CIMALVIHZVKKPE-SNAWJCMRSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
UN 3335
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?