EMAIL THIS PAGE TO A FRIEND
574775 Sigma-Aldrich

Cesium triiodide

≥99.99%

Purchase

Properties

assay   ≥99.99%
mp   207 °C (lit.)
density   4.51 g/mL at 25 °C (lit.)
SMILES string   I[Cs](I)I
InChI   1S/Cs.I3/c;1-3-2/q+1;-1
InChI key   PSDVEXXHXQVSQT-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?