• USA Home
  • 574953 - Hexaamminenickel(II) iodide

EMAIL THIS PAGE TO A FRIEND
574953 Sigma-Aldrich

Hexaamminenickel(II) iodide

99.999% trace metals basis

Purchase

Properties

assay   99.999% trace metals basis
form   powder
SMILES string   N.N.N.N.N.N.I[Ni]I
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?