• USA Home
  • 575151 - Europium(III) sulfate

EMAIL THIS PAGE TO A FRIEND
575151 Sigma-Aldrich

Europium(III) sulfate

≥99.99% trace metals basis

Synonym: Europous sulfate

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Essential Chemicals, Europium Catalysts, Europium Salts,
assay   ≥99.99% trace metals basis
form   crystalline
SMILES string   [Eu+3].[Eu+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
InChI   1S/2Eu.3H2O4S/c;;3*1-5(2,3)4/h;;3*(H2,1,2,3,4)/q2*+3;;;/p-6
InChI key   FBUHTUYBHREGEH-UHFFFAOYSA-H
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Single- & multi-channel pipettes from BRAND
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?