• USA Home
  • 575291 - Potassium trifluorohydroxyborate

EMAIL THIS PAGE TO A FRIEND
575291 Sigma-Aldrich

Potassium trifluorohydroxyborate

≥99.99% trace metals basis

Purchase

Properties

assay   ≥99.99% trace metals basis
form   powder
SMILES string   [K+].O[B-](F)(F)F
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?