• USA Home
  • 575852 - Cerium(IV)-zirconium(IV) oxide

EMAIL THIS PAGE TO A FRIEND
575852 Sigma-Aldrich

Cerium(IV)-zirconium(IV) oxide

Green Alternative

nanopowder

Purchase

Properties

form   nanopowder
composition   Zirconia, 25%
greener alternative product characteristics   Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
particle size   ≤100 nm
surface area   100-120 m2/g
density   6.61 g/mL at 25 °C (lit.)
SMILES string   O=[Zr]=O.O=[Ce]=O
InChI   1S/Ce.4O.Zr
InChI key   WKROXIWGJAIPLZ-UHFFFAOYSA-N

Description

Features and Benefits

Optically passive electrode for solid state electrochromic devices.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

634174 Cerium(IV)-zirconium(IV) oxide, nanopowder, <50 nm particle size (BET), 99.0% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?