• USA Home
  • 575917 - Dichlorotetrakis(pyridine)iron

EMAIL THIS PAGE TO A FRIEND
575917 Sigma-Aldrich

Dichlorotetrakis(pyridine)iron

Synonym: Tetrapyridineiron(III) chloride

  • CAS Number 15138-92-8

  • Empirical Formula (Hill Notation) C20H20Cl2FeN4

  • Molecular Weight 443.15

  •  MDL number MFCD10567427

  •  PubChem Substance ID 24880831

Purchase

Properties

Related Categories Electronic Chemicals, Essential Chemicals, Iron Salts, Materials Science, Metal and Ceramic Science,
InChI Key   KVQSAWIBLUAVAB-UHFFFAOYSA-L
form   powder
mp   >300 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Single- & multi-channel pipettes from BRAND
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?