• USA Home
  • 576298 - 5-(4-Methoxyphenyl)-2-furoic acid

EMAIL THIS PAGE TO A FRIEND
576298 Sigma-Aldrich

5-(4-Methoxyphenyl)-2-furoic acid

97%

Purchase

Properties

Related Categories Building Blocks, C12 to C20+, Chemical Synthesis, Furans, Heterocyclic Building Blocks More...
assay   97%
mp   182-186 °C (dec.) (lit.)
SMILES string   COc1ccc(cc1)-c2ccc(o2)C(O)=O
InChI   1S/C12H10O4/c1-15-9-4-2-8(3-5-9)10-6-7-11(16-10)12(13)14/h2-7H,1H3,(H,13,14)
InChI key   YEBLYHKPTQJGEC-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?