EMAIL THIS PAGE TO A FRIEND
576409 Sigma-Aldrich

IR-780 perchlorate

Dye content 99 %

Synonym: 1,1′,3,3,3′,3′-4,4′,5,5′-di-benzo-2,2′-indotricarbocyanine perchlorate, HDITC perchlorate

Purchase

Properties

InChI Key   RPNNJVODIWTCJW-UHFFFAOYSA-M
composition   Dye content, 99%
mp   190.6 °C (dec.) (lit.)
solubility   H2O: soluble
absorption   λmax 781 nm

Description

Packaging

500 mg in glass insert

Application

Water soluble, laser and near-IR dye

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?