• USA Home
  • 578312 - 7-Hydroxy-4-methyl-3-phenylcoumarin

EMAIL THIS PAGE TO A FRIEND
578312 Sigma-Aldrich

7-Hydroxy-4-methyl-3-phenylcoumarin

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Coumarins, Heterocyclic Building Blocks
assay   97%
mp   229-232 °C (lit.)
SMILES string   CC1=C(C(=O)Oc2cc(O)ccc12)c3ccccc3
InChI   1S/C16H12O3/c1-10-13-8-7-12(17)9-14(13)19-16(18)15(10)11-5-3-2-4-6-11/h2-9,17H,1H3
InChI key   DTDWZXAWHROHIO-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?