• USA Home
  • 578339 - 5,7-Dihydroxy-4-propylcoumarin

EMAIL THIS PAGE TO A FRIEND
578339 Sigma-Aldrich

5,7-Dihydroxy-4-propylcoumarin

98%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Coumarins, Heterocyclic Building Blocks
assay   98%
mp   239 °C (dec.) (lit.)
SMILES string   CCCC1=CC(=O)Oc2cc(O)cc(O)c12
InChI   1S/C12H12O4/c1-2-3-7-4-11(15)16-10-6-8(13)5-9(14)12(7)10/h4-6,13-14H,2-3H2,1H3
InChI key   HDPADVKOPBBPJW-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?