• USA Home
  • 58335 - (R)-(−)-Isobornylamine hydrochloride

EMAIL THIS PAGE TO A FRIEND
58335 Sigma-Aldrich

(R)-(−)-Isobornylamine hydrochloride

≥97.0% (AT)

Synonym: exo-(1R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-amine hydrochloride

  • CAS Number 24629-78-5

  • Empirical Formula (Hill Notation) C10H19N · HCl

  • Molecular Weight 189.73

  •  Beilstein/REAXYS Number 4196859

  •  EC Number 246-373-5

  •  MDL number MFCD00066428

  •  PubChem Substance ID 57651650

  •  NACRES NA.22

Purchase

Properties

Related Categories Amine Salts, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
assay   ≥97.0% (AT)
optical activity   [α]20/D −48±3°, c = 5% in ethanol
mp   ≥300 °C
SMILES string   Cl[H].[H][C@@]12CC[C@@](C)([C@H](N)C1)C2(C)C
InChI   1S/C10H19N.ClH/c1-9(2)7-4-5-10(9,3)8(11)6-7;/h7-8H,4-6,11H2,1-3H3;1H/t7-,8-,10+;/m1./s1
InChI key   XVVITZVHMWAHIG-NDYRHZEOSA-N

Description

Packaging

100 mg in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
RTECS 
ED6183800
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?