• USA Home
  • 586471 - E-α-(4-Chlorophenyl)cinnamonitrile

EMAIL THIS PAGE TO A FRIEND
586471 Sigma-Aldrich

E-α-(4-Chlorophenyl)cinnamonitrile

97%

Purchase

Properties

Related Categories Building Blocks, C10 to C27, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds - Organic Building Blocks,
assay   97%
mp   113-117 °C (lit.)
SMILES string   [H]\C(c1ccccc1)=C(\C#N)c2ccc(Cl)cc2
InChI   1S/C15H10ClN/c16-15-8-6-13(7-9-15)14(11-17)10-12-4-2-1-3-5-12/h1-10H/b14-10+
InChI key   JJGLLZWUWIKTAG-GXDHUFHOSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?