• USA Home
  • 588504 - 1-[(Trimethylsilyl)ethynyl]-3,5-dimethoxybenzene

EMAIL THIS PAGE TO A FRIEND
588504 Sigma-Aldrich

1-[(Trimethylsilyl)ethynyl]-3,5-dimethoxybenzene

97%

Synonym: 3,5-Dimethoxy-1-(trimethylsilylethynyl)benzene, [(3,5-Dimethoxyphenyl)ethynyl]trimethylsilane

Purchase

Properties

Related Categories Alkynes, Building Blocks, Chemical Synthesis, Internal, Organic Building Blocks More...
assay   97%
mp   61-65 °C (lit.)
SMILES string   COc1cc(OC)cc(c1)C#C[Si](C)(C)C
InChI   1S/C13H18O2Si/c1-14-12-8-11(6-7-16(3,4)5)9-13(10-12)15-2/h8-10H,1-5H3
InChI key   DGSBDARVIGVKSP-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles

Related Content

Alkynes and Acetylenic Building Blocks

We list over 600 alkyne building blocks, including a wide array of propargyl alcohols, acetylenic boron reagents, and halogenated substrates for cross-coupling. Additionally, many of these are termin...
Keywords: Building blocks, Coupling reactions, Cross couplings

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?