• USA Home
  • 588865 - tert-Butyldimethylsilyl bromoacetate

EMAIL THIS PAGE TO A FRIEND
588865 Sigma-Aldrich

tert-Butyldimethylsilyl bromoacetate

97%

Purchase

Properties

Related Categories Building Blocks, C8 to C9, Carbonyl Compounds, Chemical Synthesis, Esters,
assay   97%
bp   60-62 °C/0.1 mmHg (lit.)
density   1.186 g/mL at 25 °C (lit.)
SMILES string   CC(C)(C)[Si](C)(C)OC(=O)CBr
InChI   1S/C8H17BrO2Si/c1-8(2,3)12(4,5)11-7(10)6-9/h6H2,1-5H3
InChI key   LGIGKRBWJCLGHJ-UHFFFAOYSA-N

Description

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3267 8 / PGII
WGK Germany 
3
Flash Point(F) 
203 °F
Flash Point(C) 
95 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?