• USA Home
  • 590797 - PAMAM-OS-trimethoxysilyl dendrimer, generation 4 solution

EMAIL THIS PAGE TO A FRIEND
590797 Sigma-Aldrich

PAMAM-OS-trimethoxysilyl dendrimer, generation 4 solution

10 wt. % in isopropanol

  • Linear Formula [NH2(CH2)2NH2]:(G=4);dendri PAMAM((NHCH2CH2CO2(CH2)3Si(OCH3)3)128

  • Molecular Weight 43847.02

  •  MDL number MFCD04975192

Purchase

Properties

Related Categories Dendrimers, Materials Science, Nanomaterials, PAMAM Dendrimers, Trimethoxysilyl Surface Groups More...
description   trimethoxysilyl surface group
form   (10wt.% in isopropanol)
mol wt   generation 4
feature   core type ethylenediamine core (2-carbon core)
concentration   10 wt. % in isopropanol
no. Surface Groups   128
refractive index   n20/D 1.3869
bp   82.4 °C
density   0.806 g/mL at 25 °C

Description

Packaging

10 g in glass bottle

Features and Benefits

Contains 128 surface organosilicon groups.

Legal Information

Manufactured by Dendritech®, Inc.

Dendritech is a registered trademark of Dendritech, Inc.

Safety & Documentation

Safety Information

RIDADR 
UN 1219 3/PG 2
WGK Germany 
1
Flash Point(F) 
53 °F
Flash Point(C) 
12 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?