EMAIL THIS PAGE TO A FRIEND
590835 Sigma-Aldrich

Toluene-12C7

99.9 atom % 12C

Synonym: Methylbenzene-12C7

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service.
Purchase

Properties

Related Categories Alphabetical Listings, Stable Isotopes, T More...
description   13C-depleted
isotopic purity   99.9 atom % 12C
quality   13C depleted
bp   110 °C (lit.)
mp   -93 °C (lit.)
density   0.864 g/mL at 25 °C
mass shift   depleted
SMILES string   [12CH3][12c]1[12cH][12cH][12cH][12cH][12cH]1
InChI   1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3/i1+0,2+0,3+0,4+0,5+0,6+0,7+0
InChI key   YXFVVABEGXRONW-QZNDBYKSSA-N

Description

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1294 - class 3 - PG 2 - Toluene
WGK Germany 
2
Flash Point(F) 
39.2 °F
Flash Point(C) 
4 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?