• USA Home
  • 591017 - (3S)-(+)-3-(1-Methylindol-3-yl)-3-phenylpropionaldehyde

EMAIL THIS PAGE TO A FRIEND
591017 Sigma-Aldrich

(3S)-(+)-3-(1-Methylindol-3-yl)-3-phenylpropionaldehyde

97%

Synonym: (3S)-(+)-3-(1-Methylindol-3-yl)-3-phenylpropanaldehyde

Purchase

Properties

Related Categories Aldehydes, Asymmetric Synthesis, Building Blocks, C13-C60, Carbonyl Compounds,
InChI Key   IAWAPDWVVBQEPB-HNNXBMFYSA-N
assay   97%
optical activity   [α]20/D +41°, c = 1% in chloroform
optical purity   ee: 99% (HPLC)
mp   63-67 °C (lit.)

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?