• USA Home
  • 592706 - Tetraammineplatinum(II) hydrogencarbonate

EMAIL THIS PAGE TO A FRIEND
592706 Sigma-Aldrich

Tetraammineplatinum(II) hydrogencarbonate

99.99% trace metals basis

Synonym: Tetrammineplatinum hydrogen carbonate

Purchase

Properties

Related Categories Essential Chemicals, Materials Science, Metal and Ceramic Science, Platinum Salts, Research Essentials,
assay   99.99% trace metals basis
form   powder
mp   236 °C (dec.) (lit.)
SMILES string   N.N.N.N.OC(=O)O[Pt]OC(O)=O
InChI   1S/2CH2O3.4H3N.Pt/c2*2-1(3)4;;;;;/h2*(H2,2,3,4);4*1H3;/q;;;;;;+2/p-2
InChI key   IXGOCCWMKLVVSM-UHFFFAOYSA-L

Description

Physical form

Macro-agglomeration of ultra-fine particles, with size about 100 nanometers.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1479 5.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Single- & multi-channel pipettes from BRAND
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?