• USA Home
  • 593222 - 3,5-Di-tert-butylphenyl trifluoromethanesulfonate

EMAIL THIS PAGE TO A FRIEND
593222 Sigma-Aldrich

3,5-Di-tert-butylphenyl trifluoromethanesulfonate

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Organic Triflates, Sulfur Compounds More...
InChI Key   NMKZEDAODGHHEC-UHFFFAOYSA-N
assay   97%
mp   24-29 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?