• USA Home
  • 593389 - 1,2-Dibromo-4-tert-butylbenzene

EMAIL THIS PAGE TO A FRIEND
593389 Sigma-Aldrich

1,2-Dibromo-4-tert-butylbenzene

97%

Purchase

Properties

assay   97%
form   solid
refractive index   n20/D 1.5710 (lit.)
bp   281-282 °C (lit.)
density   1.557 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?