• USA Home
  • 594113 - 2-Chlorophenyl isocyanide

EMAIL THIS PAGE TO A FRIEND
594113 Sigma-Aldrich

2-Chlorophenyl isocyanide

97%

Purchase

Properties

assay   97%
mp   28-32 °C (lit.)
SMILES string   Clc1ccccc1[N+]#[C-]
Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
200 °F
Flash Point(C) 
93 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?