• USA Home
  • 594210 - 2,2′,4-Trimethoxybenzophenone

EMAIL THIS PAGE TO A FRIEND
594210 Sigma-Aldrich

2,2′,4-Trimethoxybenzophenone

97%

Purchase

Properties

Related Categories Building Blocks, C15 to C38, Carbonyl Compounds, Chemical Synthesis, Ketones,
assay   97%
mp   55.5-59.5 °C (lit.)
SMILES string   COc1ccc(C(=O)c2ccccc2OC)c(OC)c1
InChI   1S/C16H16O4/c1-18-11-8-9-13(15(10-11)20-3)16(17)12-6-4-5-7-14(12)19-2/h4-10H,1-3H3
InChI key   FUGHBBQOPVKADC-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?